Gene Transcripts

Gene Symbol
Slc15a2
Entrez Gene ID
60577
Gene Name
solute carrier family 15 (oligopeptide transporter), member 2
Gene Aliases
Chromosome Location
Chr. 11: 66932664 - 66961708 on Build Rnor_6.0
UniGene ID
Rn.89268
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031672.2 NP_113860.2 2 348
GenBank mRNA BC085327.1 - 2 348
D63149.1 - 2 336

View Full Product Details