Gene Transcripts

Gene Symbol
Psmd9
Entrez Gene ID
161475
Gene Name
proteasome (prosome, macropain) 26S subunit, non-ATPase, 9
Gene Aliases
Bridge, Bridge-1
Chromosome Location
Chr. 12: 38786539 - 38805121 on Build Rnor_6.0
UniGene ID
Rn.24127
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_130430.1 NP_569114.1 6 1343
GenBank mRNA AF067728.1 - 6 1343

View Full Product Details