Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_012180.2 | NP_036312.2 | 2 | 1006 | |
XM_017008020.1 | XP_016863509.1 | 1 | 143 | ||
GenBank mRNA | AF115514.1 | - | 2 | 389 | |
AF174596.1 | - | 2 | 222 | ||
AF201932.1 | - | 2 | 415 | ||
AF233224.1 | - | 2 | 261 | ||
AK026319.1 | - | 2 | 429 | ||
AK123740.1 | - | 2 | 1019 | ||
AK314483.1 | - | 2 | 424 | ||
AL602916.1 | - | 2 | 421 | ||
AY358721.1 | - | 2 | 392 | ||
BC014679.2 | - | 2 | 408 | 3857760 | |
BC014903.1 | - | 2 | 1006 | ||
KU178588.1 | - | 1 | 156 | ||
KU178589.1 | - | 1 | 156 |