Gene Transcripts

Gene Symbol
IFIT5
Entrez Gene ID
24138
Gene Name
interferon induced protein with tetratricopeptide repeats 5
Gene Aliases
ISG58, P58, RI58
Chromosome Location
Chr. 10: 89414568 - 89421002 on Build GRCh38
UniGene ID
Hs.252839
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012420.2 NP_036552.1 2 338
GenBank mRNA AK293346.1 - 2 144
AK312358.1 - 2 171
BC025786.1 - 2 317 5229306
CR457031.1 - 1 107
DC392439.1 - 2 338
U34605.1 - 2 224

View Full Product Details