Gene Transcripts
- Gene Symbol
- Dlg1
- Entrez Gene ID
- 25252
- Gene Name
- discs, large homolog 1 (Drosophila)
- Gene Aliases
- Dlgh1, SAP97
- Chromosome Location
- Chr. 11: 72164566 - 72378895 on Build Rnor_6.0
- UniGene ID
- Rn.89331
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_012788.1 |
NP_036920.1
|
3 |
527 |
|
GenBank mRNA |
U14950.1 |
- |
3 |
527 |
|
View Full Product Details