Gene Transcripts

Gene Symbol
Tmem27
Entrez Gene ID
57395
Gene Name
transmembrane protein 27
Gene Aliases
NX-17, Nx17, collectrin
Chromosome Location
Chr. X: 32118082 - 32153687 on Build Rnor_6.0
UniGene ID
Rn.32298
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020976.1 NP_066125.1 2, 3 128
GenBank mRNA AF178086.1 - 2, 3 128
BC078838.1 - 2, 3 185 7129929

View Full Product Details