Gene Transcripts

Gene Symbol
TMEM14A
Entrez Gene ID
28978
Gene Name
transmembrane protein 14A
Gene Aliases
C6orf73, PTD011
Chromosome Location
Chr. 6: 52671086 - 52686588 on Build GRCh38
UniGene ID
Hs.94896
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014051.3 NP_054770.1 2, 3 254
GenBank mRNA AF078864.1 - 2, 3 298
AF239771.1 - 2, 3 233
AK312063.1 - 2, 3 227
BC015097.1 - 2, 3 213 3686196
BC019328.1 - 2, 3 209 3534851
CB117714.1 - 2, 3 254

View Full Product Details