Gene Transcripts

Gene Symbol
SMYD2
Entrez Gene ID
56950
Gene Name
SET and MYND domain containing 2
Gene Aliases
HSKM-B, KMT3C, ZMYND14
Chromosome Location
Chr. 1: 214281101 - 214337134 on Build GRCh38
UniGene ID
Hs.66170
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020197.2 NP_064582.2 4, 5 437
GenBank mRNA AF226053.1 - 4, 5 426
AK313868.1 - 4, 5 407
BC028104.1 - 3, 4 358
BC068511.1 - 3, 4 503
BC098133.1 - 4, 5 426 40005281
BC098276.1 - 4, 5 426 40005283
BC098305.1 - 4, 5 426 40005284
BC098335.1 - 4, 5 426 40005280

View Full Product Details