Gene Transcripts

Gene Symbol
SH3PXD2A
Entrez Gene ID
9644
Gene Name
SH3 and PX domains 2A
Gene Aliases
FISH, SH3MD1, TKS5
Chromosome Location
Chr. 10: 103594027 - 103855464 on Build GRCh38
UniGene ID
Hs.678727
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014631.2 NP_055446.2 9, 10 868
XM_005270294.4 XP_005270351.1 10, 11 1001
XM_005270295.4 XP_005270352.1 9, 10 955
XM_005270297.3 XP_005270354.1 4, 5 2232
XM_005270298.4 XP_005270355.1 4, 5 1035
XM_006718079.3 XP_006718142.1 11, 12 1098
XM_006718080.3 XP_006718143.1 5, 6 1129
XM_011540385.2 XP_011538687.1 10, 11 1052
XM_011540386.2 XP_011538688.1 5, 6 2328
XM_011540387.2 XP_011538689.1 1, 2 3974
XM_017016969.1 XP_016872458.1 10, 11 1019
XM_017016970.1 XP_016872459.1 9, 10 835
GenBank mRNA AB007878.1 - 7, 8 618
AK056469.1 - 8, 9 591
AK294454.1 - 4, 5 408
AK294736.1 - 4, 5 446
AK295314.1 - 4, 5 485
AK295660.1 - 4, 5 512
AK302015.1 - 7, 8 624
AK304828.1 - 4, 5 424
AK315983.1 - 4, 5 451

View Full Product Details