Gene Transcripts

Gene Symbol
OR10A6
Entrez Gene ID
390093
Gene Name
olfactory receptor family 10 subfamily A member 6 (gene/pseudogene)
Gene Aliases
OR11-96
Chromosome Location
Chr. 11: 7927718 - 7928662 on Build GRCh38
UniGene ID
Hs.528593
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004461.1 NP_001004461.1 1 573

View Full Product Details