Gene Transcripts

Gene Symbol
EDEM1
Entrez Gene ID
9695
Gene Name
ER degradation enhancing alpha-mannosidase like protein 1
Gene Aliases
EDEM
Chromosome Location
Chr. 3: 5187674 - 5219965 on Build GRCh38
UniGene ID
Hs.224616
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014674.2 NP_055489.1 5, 6 1173
XM_011534271.2 XP_011532573.1 5, 6 1367
XM_011534272.2 XP_011532574.1 6, 7 1166
GenBank mRNA AK292643.1 - 5, 6 1041
AK298511.1 - 5, 6 601
AK302065.1 - 5, 6 642
BC019088.1 - 5, 6 1180 4863779
D86967.1 - 5, 6 1099

View Full Product Details