Gene Transcripts

Gene Symbol
Klk10
Entrez Gene ID
292850
Gene Name
kallikrein related-peptidase 10
Gene Aliases
Chromosome Location
Chr. 1: 99718433 - 99722595 on Build Rnor_6.0
UniGene ID
Rn.230292
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004100.1 NP_001004100.1 3 422
XM_006228987.2 XP_006229049.1 4 557

View Full Product Details