Gene Transcripts

Gene Symbol
Omg
Entrez Gene ID
450224
Gene Name
oligodendrocyte-myelin glycoprotein
Gene Aliases
Chromosome Location
Chr. 10: 66845654 - 66848388 on Build Rnor_6.0
UniGene ID
Rn.163139
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005898.3 NP_001005898.3 2 616
GenBank mRNA AY250703.1 - 2 590
AY316298.1 - 1 464
BC087723.1 - 1 482 7320740

View Full Product Details