Gene Transcripts

Gene Symbol
Gc
Entrez Gene ID
14473
Gene Name
group specific component
Gene Aliases
DBP, VDB
Chromosome Location
Chr. 5: 89417511 - 89457898 on Build GRCm38
UniGene ID
Mm.196595
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008096.2 NP_032122.1 9 1286
GenBank mRNA AK010965.1 - 9 1110
BC010762.1 - 9 1149 4234788
BC051395.1 - 9 1153 5125043
M55413.1 - 9 1096

View Full Product Details