Gene Transcripts

Gene Symbol
SUMF2
Entrez Gene ID
25870
Gene Name
sulfatase modifying factor 2
Gene Aliases
pFGE
Chromosome Location
Chr. 7: 56064007 - 56080672 on Build GRCh38
UniGene ID
Hs.279696
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001042469.1 NP_001035934.2 2 174
NM_001042470.1 NP_001035935.2 2 174
NM_001130069.2 NP_001123541.1 2 174
NM_015411.2 NP_056226.2 2 174
XM_011515254.2 XP_011513556.1 2 174
XM_017011939.1 XP_016867428.1 2 174
XM_017011940.1 XP_016867429.1 2 391
GenBank mRNA AK075477.1 - 2 133
AK297042.1 - 2 90
AK297793.1 - 2 88
AK298605.1 - 2 114
AK301627.1 - 2 91
AL050037.1 - 2 90
AY323911.1 - 2 86
AY359103.1 - 2 104
BC006159.1 - 2 72
BC015600.2 - 2 71
BC065222.1 - 2 121
BC084539.1 - 2 104 6599080
BC111092.1 - 2 127 40068651
BM423367.1 - 2 76
BP296294.1 - 2 396
CR936757.1 - 2 117
CX164099.1 - 2 52
DA130450.1 - 2 143
DA917807.1 - 2 143
DA924174.1 - 2 175
DB010869.1 - 2 92

View Full Product Details