Gene Transcripts

Gene Symbol
ACIN1
Entrez Gene ID
22985
Gene Name
apoptotic chromatin condensation inducer 1
Gene Aliases
ACINUS, ACN, fSAP152
Chromosome Location
Chr. 14: 23058564 - 23095614 on Build GRCh38
UniGene ID
Hs.124490
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164815.1 NP_001158287.1 10, 11 2819
NM_001164816.1 NP_001158288.1 4, 5 458
NM_001164817.1 NP_001158289.1 5, 6 520
NM_014977.3 NP_055792.1 11, 12 2939
XM_005267415.3 XP_005267472.1 11, 12 2740
XM_005267418.1 XP_005267475.1 4, 5 755
GenBank mRNA AB014570.2 - 11, 12 2448
AF124726.1 - 11, 12 2938
AF124727.1 - 5, 6 474
AF124728.1 - 4, 5 449
AK299034.1 - 10, 11 2330
AL833228.1 - 6, 7 789
BX247975.1 - 4, 5 728

View Full Product Details