Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001204477.1 | NP_001191406.1 | 4 | 385 | |
GenBank mRNA | AK093993.1 | - | 2 | 129 | |
BC029542.1 | - | 4 | 385 | 5298701 | |
BX379636.2 | - | 2 | 152 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001204478.1 | NP_001191407.1 | 7 | 796 | |
NR_037924.1 | 6 | 651 | |||
GenBank mRNA | AB074181.1 | - | 7 | 726 | |
AK303475.1 | - | 7 | 765 | ||
DA796743.1 | - | 6 | 492 |