Gene Transcripts

Gene Symbol
MOK
Entrez Gene ID
5891
Gene Name
MOK protein kinase
Gene Aliases
RAGE, RAGE-1, RAGE1, STK30
Chromosome Location
Chr. 14: 102224450 - 102305200 on Build GRCh38
UniGene ID
Hs.104119
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001272011.1 NP_001258940.1 2 308
NM_014226.2 NP_055041.1 2 308
XM_006720225.2 XP_006720288.1 2 308
XM_006720226.2 XP_006720289.1 2 308
XM_011537052.1 XP_011535354.1 2 308
XM_017021549.1 XP_016877038.1 2 308
XM_017021550.1 XP_016877039.1 2 297
XM_017021551.1 XP_016877040.1 2 213
XM_017021552.1 XP_016877041.1 2 262
XM_017021553.1 XP_016877042.1 2 307
XM_017021554.1 XP_016877043.1 2 303
XM_017021555.1 XP_016877044.1 2 308
XM_017021556.1 XP_016877045.1 2 308
XM_017021557.1 XP_016877046.1 2 298
XM_017021558.1 XP_016877047.1 2 264
XM_017021559.1 XP_016877048.1 2 294
GenBank mRNA AB022694.1 - 2 302
AK094747.1 - 2 276
AK302349.1 - 2 293
AK312778.1 - 2 299
BC026069.1 - 2 298 4838296
BC053536.1 - 2 284 5175851
DB037795.1 - 2 308

View Full Product Details