Gene Transcripts
- Gene Symbol
- MOK
- Entrez Gene ID
- 5891
- Gene Name
- MOK protein kinase
- Gene Aliases
- RAGE, RAGE-1, RAGE1, STK30
- Chromosome Location
- Chr. 14: 102224450 - 102305200 on Build GRCh38
- UniGene ID
- Hs.104119
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001272011.1 |
NP_001258940.1
|
5, 6 |
564 |
|
NM_014226.2 |
NP_055041.1
|
6, 7 |
654 |
|
XM_006720225.2 |
XP_006720288.1
|
6, 7 |
654 |
|
XM_006720226.2 |
XP_006720289.1
|
6, 7 |
654 |
|
XM_011537052.1 |
XP_011535354.1
|
6, 7 |
654 |
|
XM_017021551.1 |
XP_016877040.1
|
7, 8 |
594 |
|
XM_017021554.1 |
XP_016877043.1
|
4, 5 |
488 |
|
XM_017021556.1 |
XP_016877045.1
|
6, 7 |
654 |
|
XM_017021557.1 |
XP_016877046.1
|
5, 6 |
554 |
|
XM_017021559.1 |
XP_016877048.1
|
5, 6 |
643 |
|
GenBank mRNA |
AB022694.1 |
- |
6, 7 |
648 |
|
AK094747.1 |
- |
4, 5 |
461 |
|
AK302349.1 |
- |
5, 6 |
549 |
|
AK312778.1 |
- |
6, 7 |
645 |
|
BC026069.1 |
- |
5, 6 |
1065 |
4838296 |
BC043179.1 |
- |
4, 5 |
1410 |
5287879 |
BC053536.1 |
- |
6, 7 |
630 |
5175851 |
View Full Product Details