Gene Transcripts

Gene Symbol
HAUS4
Entrez Gene ID
54930
Gene Name
HAUS augmin like complex subunit 4
Gene Aliases
C14orf94
Chromosome Location
Chr. 14: 22946228 - 22957142 on Build GRCh38
UniGene ID
Hs.442782
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166269.1 NP_001159741.1 3 353
NM_001166270.1 NP_001159742.1 3 345
NM_017815.2 NP_060285.2 3 345
GenBank mRNA AK000431.1 - 3 314
AK026016.1 - 3 299
AK057779.1 - 3 421
BC001916.1 - 3 342 2905474
BC002554.2 - 3 276 3139198
BQ066232.1 - 3 353
BX247981.1 - 3 529
BX248256.1 - 3 312

View Full Product Details