Gene Transcripts

Gene Symbol
COA6
Entrez Gene ID
388753
Gene Name
cytochrome c oxidase assembly factor 6
Gene Aliases
C1orf31, CEMCOX4
Chromosome Location
Chr. 1: 234373437 - 234384049 on Build GRCh38
UniGene ID
Hs.23198
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012985.2 NP_001013003.1 3 503
NM_001206641.2 NP_001193570.2 3 587
NM_001301733.1 NP_001288662.1 2 874
GenBank mRNA BC025793.1 - 3 410
BC035925.1 - 3 500
BC116455.1 - 3 503 4540930
BG195363.1 - 1 190
BG204581.1 - 2 346
CB136291.1 - 3 556

View Full Product Details