Gene Transcripts

Gene Symbol
LOC500227
Entrez Gene ID
500227
Gene Name
hypothetical gene supported by BC079424
Gene Aliases
Chromosome Location
Chr. 4: 114854449 - 114860678 on Build Rnor_6.0
UniGene ID
Rn.50288
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024330.1 NP_001019501.1 4 548
XM_006236784.2 XP_006236846.1 3 335
GenBank mRNA BC079424.1 - 4 548

View Full Product Details