Gene Transcripts

Gene Symbol
Bola1
Entrez Gene ID
69168
Gene Name
bolA-like 1 (E. coli)
Gene Aliases
1810037G04Rik
Chromosome Location
Chr. 3: 96196583 - 96200408 on Build GRCm38
UniGene ID
Mm.104531
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026975.2 NP_081251.1 2 484
GenBank mRNA AK007718.1 - 2 485
BC027558.1 - 2 480 1477943

View Full Product Details