Gene Transcripts

Gene Symbol
Fgf4
Entrez Gene ID
14175
Gene Name
fibroblast growth factor 4
Gene Aliases
Fgf-4, Fgfk, Hst1, Hstf-1, KS3, hst, hst-1, kFGF
Chromosome Location
Chr. 7: 144861386 - 144865243 on Build GRCm38
UniGene ID
Mm.4956
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010202.5 NP_034332.2 3 1862
GenBank mRNA AK082814.1 - 3 1863

View Full Product Details