Gene Transcripts

Gene Symbol
KIAA0930
Entrez Gene ID
23313
Gene Name
KIAA0930
Gene Aliases
C22orf9, LSC3
Chromosome Location
Chr. 22: 45192242 - 45240769 on Build GRCh38
UniGene ID
Hs.592207
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009880.1 NP_001009880.1 10 1830
NM_015264.1 NP_056079.1 10 1902
XM_011530017.2 XP_011528319.1 10 1865
XM_017028672.1 XP_016884161.1 10 1874
GenBank mRNA AB023147.1 - 9 1598
AK025608.1 - 8 1419
AK094359.1 - 1 1356
AK094937.1 - 11 1913
AK124648.1 - 10 1910
AY007105.1 - 3 784
BC016882.1 - 5 1160
BC038414.1 - 9 1977
BC053595.1 - 10 1824 5730020

View Full Product Details