Gene Transcripts

Gene Symbol
ALG1L
Entrez Gene ID
200810
Gene Name
ALG1, chitobiosyldiphosphodolichol beta-mannosyltransferase like
Gene Aliases
ALG1L1
Chromosome Location
Chr. 3: 125929272 - 125993832 on Build GRCh38
UniGene ID
Hs.591299
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001015050.2 NP_001015050.2 6 556
NM_001195223.1 NP_001182152.1 7 667
XM_017005844.1 XP_016861333.1 7 3934
XM_017005845.1 XP_016861334.1 7 560
XM_017005846.1 XP_016861335.1 6 3823
XM_017005847.1 XP_016861336.1 6 449
GenBank mRNA BC073816.1 - 6 551
BI085096.1 - 7 660
CD685738.1 - 6 577

View Full Product Details