Gene Transcripts

Gene Symbol
TMEM35A
Entrez Gene ID
59353
Gene Name
transmembrane protein 35A
Gene Aliases
TMEM35
Chromosome Location
Chr. X: 101078847 - 101096367 on Build GRCh38
UniGene ID
Hs.45140
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021637.2 NP_067650.1 2 1824
GenBank mRNA AK024146.1 - 2 1795
AK223240.1 - 2 1791
BC078658.1 - 2 1788 5261160

View Full Product Details