Gene Transcripts

Gene Symbol
ARHGEF28
Entrez Gene ID
64283
Gene Name
Rho guanine nucleotide exchange factor 28
Gene Aliases
RGNEF, RIP2, p190RHOGEF
Chromosome Location
Chr. 5: 73626158 - 73941993 on Build GRCh38
UniGene ID
Hs.482521
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080479.2 NP_001073948.2 25 3418
NM_001177693.1 NP_001171164.1 25 3418
NM_001244364.1 NP_001231293.1 17 2374
XM_005248568.4 XP_005248625.1 17 3855
XM_011543546.1 XP_011541848.1 25 3425
XM_011543547.2 XP_011541849.1 24 3652
XM_017009702.1 XP_016865191.1 24 3103
XM_017009703.1 XP_016865192.1 16 2156
GenBank mRNA AK025470.1 - 1 134
AK025816.1 - 2 314
AK304761.1 - 17 2374
BC012946.1 - 2 292
BC157846.1 - 24 3264 9057065
BC171850.1 - 24 3264 9054504
CR936816.1 - 3 421

View Full Product Details