Gene Transcripts

Gene Symbol
NAA50
Entrez Gene ID
80218
Gene Name
N(alpha)-acetyltransferase 50, NatE catalytic subunit
Gene Aliases
MAK3, NAT13, NAT13P, NAT5, NAT5P, SAN
Chromosome Location
Chr. 3: 113716460 - 113746299 on Build GRCh38
UniGene ID
Hs.372378
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001308445.1 NP_001295374.1 5 1665
NM_025146.3 NP_079422.1 5 1668
GenBank mRNA AK022540.1 - 1 974
AK023090.1 - 5 1582
AK023256.1 - 5 1618
AK296456.1 - 5 1363
BC012731.2 - 5 1607 3619903
CR749314.1 - 3 1299

View Full Product Details