Gene Transcripts
- Gene Symbol
- ATP10B
- Entrez Gene ID
- 23120
- Gene Name
- ATPase phospholipid transporting 10B (putative)
- Gene Aliases
- ATPVB
- Chromosome Location
- Chr. 5: 160563120 - 160929411 on Build GRCh38
- UniGene ID
- Hs.109358
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK025130.1 |
- |
7 |
1549 |
|
AK090832.1 |
- |
11 |
2256 |
|
AK295081.1 |
- |
10 |
2106 |
|
View Full Product Details