Gene Transcripts

Gene Symbol
FAM49A
Entrez Gene ID
81553
Gene Name
family with sequence similarity 49 member A
Gene Aliases
Chromosome Location
Chr. 2: 16549459 - 16665866 on Build GRCh38
UniGene ID
Hs.467769
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030797.3 NP_110424.1 6 648
XM_005262630.3 XP_005262687.1 5 604
XM_006712108.3 XP_006712171.1 6 603
GenBank mRNA AK055334.1 - 6 616
AK091280.1 - 1 62
AK315065.1 - 5 543
AL136704.1 - 6 613
BC038971.1 - 6 599 5200807
CR533504.1 - 4 395

View Full Product Details