Gene Transcripts

Gene Symbol
SH2D1B
Entrez Gene ID
117157
Gene Name
SH2 domain containing 1B
Gene Aliases
EAT2
Chromosome Location
Chr. 1: 162395266 - 162412138 on Build GRCh38
UniGene ID
Hs.350581
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053282.4 NP_444512.2 3 338
GenBank mRNA AF256653.1 - 3 216
AF403479.1 - 3 216
AF484964.1 - 3 314
AK097162.1 - 3 265
AK314743.1 - 3 285
BC022407.1 - 3 326 4717041

View Full Product Details