Gene Transcripts

Gene Symbol
SPATA17
Entrez Gene ID
128153
Gene Name
spermatogenesis associated 17
Gene Aliases
IQCH, MSRG-11, MSRG11
Chromosome Location
Chr. 1: 217631324 - 217868151 on Build GRCh38
UniGene ID
Hs.171130
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138796.3 NP_620151.1 2 156
XM_005273052.1 XP_005273109.1 2 138
XM_006711165.3 XP_006711228.1 2 147
XM_011509194.2 XP_011507496.1 2 138
GenBank mRNA AK098591.1 - 2 136
AK310291.1 - 2 140
AY963797.1 - 2 159
BC014608.2 - 2 127 4824429

View Full Product Details