Gene Transcripts

Gene Symbol
FAM69B
Entrez Gene ID
138311
Gene Name
family with sequence similarity 69 member B
Gene Aliases
C9orf136, pp6977
Chromosome Location
Chr. 9: 136712572 - 136724718 on Build GRCh38
UniGene ID
Hs.741305
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152421.3 NP_689634.2 4 435
XM_006716954.3 XP_006717017.1 4 440
XM_011518238.2 XP_011516540.1 5 566
XM_017014273.1 XP_016869762.1 3 473
GenBank mRNA AF318355.1 - 2 1180
AJ420457.1 - 3 275
BC032097.2 - 4 429 3161575
BG700129.1 - 4 440

View Full Product Details