Gene Transcripts

Gene Symbol
ZNF75A
Entrez Gene ID
7627
Gene Name
zinc finger protein 75a
Gene Aliases
Chromosome Location
Chr. 16: 3305406 - 3322999 on Build GRCh38
UniGene ID
Hs.513292
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001302109.1 NP_001289038.1 7 1419
NM_001324041.1 NP_001310970.1 7 1376
NM_001324085.1 NP_001311014.1 5 1112
NM_153028.3 NP_694573.1 6 895
NR_126069.1 5 1054
NR_136681.1 6 1223
NR_136683.1 5 699
NR_136686.1 6 1249
XM_011522646.2 XP_011520948.1 6 1308
XM_011522647.2 XP_011520949.1 6 993
XM_017023642.1 XP_016879131.1 6 791
GenBank mRNA AK056091.1 - 6 818
AK093979.1 - 5 1007
BC119652.1 - 5 620 40115880
BC119653.1 - 5 620 40115881
CR749782.1 - 6 718

View Full Product Details