Gene Transcripts

Gene Symbol
SNX20
Entrez Gene ID
124460
Gene Name
sorting nexin 20
Gene Aliases
SLIC1
Chromosome Location
Chr. 16: 50666300 - 50681353 on Build GRCh38
UniGene ID
Hs.460777
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153337.2 NP_699168.1 4 1334
GenBank mRNA AK055837.1 - 4 1299
BC027944.1 - 4 1283

View Full Product Details