Gene Transcripts

Gene Symbol
IGSF22
Entrez Gene ID
283284
Gene Name
immunoglobulin superfamily member 22
Gene Aliases
IGFN2
Chromosome Location
Chr. 11: 18704305 - 18726939 on Build GRCh38
UniGene ID
Hs.434152
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173588.3 NP_775859.3 3 323
XM_011520030.2 XP_011518332.1 1 3227
XM_011520031.2 XP_011518333.1 1 3227
XM_017017607.1 XP_016873096.1 2 3118
XM_017017608.1 XP_016873097.1 2 1141
GenBank mRNA AK095113.1 - 3 323
BC115396.1 - 3 274
BC115397.1 - 3 274 40068579
DA338373.1 - 3 281

View Full Product Details