Gene Transcripts

Gene Symbol
IGSF22
Entrez Gene ID
283284
Gene Name
immunoglobulin superfamily member 22
Gene Aliases
IGFN2
Chromosome Location
Chr. 11: 18704305 - 18726939 on Build GRCh38
UniGene ID
Hs.434152
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173588.3 NP_775859.3 5, 6 676
XM_011520030.2 XP_011518332.1 3, 4 3580
XM_011520031.2 XP_011518333.1 3, 4 3580
XM_017017607.1 XP_016873096.1 4, 5 3471
XM_017017608.1 XP_016873097.1 4, 5 1494
GenBank mRNA AK095113.1 - 5, 6 676
BC115396.1 - 5, 6 627
BC115397.1 - 5, 6 627 40068579

View Full Product Details