Gene Transcripts

Gene Symbol
FXYD7
Entrez Gene ID
53822
Gene Name
FXYD domain containing ion transport regulator 7
Gene Aliases
Chromosome Location
Chr. 19: 35143250 - 35154302 on Build GRCh38
UniGene ID
Hs.134729
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022006.1 NP_071289.1 4 233
GenBank mRNA AK057825.1 - 4 208
BC018619.1 - 4 217 3626060

View Full Product Details