Gene Transcripts

Gene Symbol
Parp1
Entrez Gene ID
25591
Gene Name
poly (ADP-ribose) polymerase 1
Gene Aliases
ARTD1, Adprt, Parp-1
Chromosome Location
Chr. 13: 98857255 - 98889444 on Build Rnor_6.0
UniGene ID
Rn.11327
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013063.2 NP_037195.1 3 448
GenBank mRNA BC085765.1 - 3 448
U94340.1 - 3 416

View Full Product Details