Gene Transcripts

Gene Symbol
Pdlim4
Entrez Gene ID
24915
Gene Name
PDZ and LIM domain 4
Gene Aliases
H-Rev18, Ril
Chromosome Location
Chr. 10: 39390578 - 39405311 on Build Rnor_6.0
UniGene ID
Rn.34221
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017062.2 NP_058758.1 2 288
XM_006246230.2 XP_006246292.1 2 633
XM_006246231.2 XP_006246293.1 2 346
GenBank mRNA X76454.1 - 2 289

View Full Product Details