Gene Transcripts

Gene Symbol
Aadat
Entrez Gene ID
29416
Gene Name
aminoadipate aminotransferase
Gene Aliases
Kat2
Chromosome Location
Chr. 16: 32832001 - 32868680 on Build Rnor_6.0
UniGene ID
Rn.11133
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017193.1 NP_058889.1 2 313
XM_006253067.2 XP_006253129.1 2 401
GenBank mRNA BC078864.1 - 2 309
Z50144.1 - 2 313

View Full Product Details