Gene Transcripts

Gene Symbol
Oxsr1
Entrez Gene ID
108737
Gene Name
oxidative-stress responsive 1
Gene Aliases
2210022N24Rik, 2810422B09Rik, AI462649, AW209236, Osr1, mKIAA1101
Chromosome Location
Chr. 9: 119238432 - 119322710 on Build GRCm38
UniGene ID
Mm.293565
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133985.2 NP_598746.2 7, 8 1056
GenBank mRNA AK031790.1 - 6, 7 553
AK075837.1 - 6, 7 564
AK129291.1 - 7, 8 1051
BC046453.1 - 8, 9 1080
BC060645.1 - 7, 8 1039 6852021

View Full Product Details