Gene Transcripts

Gene Symbol
Camsap1
Entrez Gene ID
227634
Gene Name
calmodulin regulated spectrin-associated protein 1
Gene Aliases
9530003A05Rik, C77823, PRO2405
Chromosome Location
Chr. 2: 25926838 - 25983452 on Build GRCm38
UniGene ID
Mm.36834
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001276359.1 NP_001263288.1 16 4774
NM_001276360.1 NP_001263289.1 17 4807
NM_001276361.1 NP_001263290.1 17 4834
GenBank mRNA AK035221.1 - 9 3286
AK170150.1 - 6 1439

View Full Product Details