Gene Transcripts

Gene Symbol
Greb1l
Entrez Gene ID
381157
Gene Name
growth regulation by estrogen in breast cancer-like
Gene Aliases
mKIAA4095
Chromosome Location
Chr. 18: 10324963 - 10562941 on Build GRCm38
UniGene ID
Mm.261535
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001083628.1 NP_001077097.1 33 7755
GenBank mRNA AK220484.1 - 13 4272

View Full Product Details