Gene Transcripts

Gene Symbol
Wt1
Entrez Gene ID
24883
Gene Name
Wilms tumor 1
Gene Aliases
Chromosome Location
Chr. 3: 95133242 - 95180574 on Build Rnor_6.0
UniGene ID
Rn.92531
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031534.2 NP_113722.2 9, 10 1239
XM_006234620.1 XP_006234682.1 8, 9 1581
GenBank mRNA CO382418.1 - 1, 2 107
X69716.1 - 9, 10 1239

View Full Product Details