Gene Transcripts

Gene Symbol
Pnliprp1
Entrez Gene ID
84028
Gene Name
pancreatic lipase-related protein 1
Gene Aliases
Chromosome Location
Chr. 1: 279867034 - 279882663 on Build Rnor_6.0
UniGene ID
Rn.6696
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032081.1 NP_114470.1 9, 10 1001
GenBank mRNA FQ232772.1 - 9, 10 1001
X61925.1 - 9, 10 1001

View Full Product Details