Gene Transcripts

Gene Symbol
Hdlbp
Entrez Gene ID
64474
Gene Name
high density lipoprotein binding protein
Gene Aliases
Chromosome Location
Chr. 9: 100554574 - 100624638 on Build Rnor_6.0
UniGene ID
Rn.8515
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172039.2 NP_742036.2 13 1714
XM_006245548.1 XP_006245610.1 13 1754
GenBank mRNA BC105748.1 - 13 1714
U90725.1 - 13 1714

View Full Product Details