Gene Transcripts

Gene Symbol
Pias2
Entrez Gene ID
17344
Gene Name
protein inhibitor of activated STAT 2
Gene Aliases
6330408K17Rik, AI462206, ARIP3, AU018068, Dib, Miz1, PIASxalpha, PIASxb, PIASxbeta, SIZ2
Chromosome Location
Chr. 18: 77065208 - 77155710 on Build GRCm38
UniGene ID
Mm.6370
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164167.1 NP_001157639.1 7 1130
NM_001164168.1 NP_001157640.1 7 1146
NM_001164169.1 NP_001157641.1 7 1130
NM_001164170.1 NP_001157642.1 7 1146
NM_008602.4 NP_032628.3 7 1130
GenBank mRNA AF039567.1 - 6 575
AF201391.1 - 7 1057
AK014871.1 - 7 1121
AK029716.1 - 7 1131
AK078097.1 - 2 198
AK078813.1 - 7 1100
AK086653.1 - 7 1062
BC005596.1 - 7 1007 3969546
BC034711.1 - 7 1136 2631738

View Full Product Details