Gene Transcripts

Gene Symbol
MIXL1
Entrez Gene ID
83881
Gene Name
Mix paired-like homeobox
Gene Aliases
MILD1, MIX, MIXL
Chromosome Location
Chr. 1: 226223618 - 226227055 on Build GRCh38
UniGene ID
Hs.282079
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282402.1 NP_001269331.1 2 489
NM_031944.2 NP_114150.1 1, 2 465
GenBank mRNA AF211891.1 - 1, 2 401
BC111974.1 - 1, 2 465 8327442
BC113441.1 - 1, 2 465 8322493
BC143784.1 - 2 489 9052298

View Full Product Details